Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290818.1 | NP_001277747.1 | 14 - 15 | 1437 | 74 | |
NM_130863.2 | NP_570933.1 | 15 - 16 | 1509 | 74 | ||
GenBank mRNA | AF333028.1 | - | 14 - 15 | 1229 | 74 | |
AK155691.1 | - | 14 - 15 | 1438 | 74 | ||
BC003196.1 | - | 14 - 15 | 1225 | 74 | ||
BC033272.1 | - | 13 - 14 | 1553 | 74 | ||
BC053922.1 | - | 15 - 16 | 1242 | 6309555 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |