Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172468.2 | NP_766056.1 | 3 - 4 | 885 | 57 | |
GenBank mRNA | AK029012.1 | - | 3 - 4 | 687 | 57 | |
AK147437.1 | - | 3 - 4 | 887 | 57 | ||
AK165619.1 | - | 3 - 4 | 695 | 57 | ||
AK170774.1 | - | 3 - 4 | 659 | 57 | ||
AK170999.1 | - | 3 - 4 | 687 | 57 | ||
BC099674.1 | - | 3 - 4 | 681 | 4950374 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |