Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177320.2 | NP_796294.2 | 15 - 16 | 2337 | 108 | |
GenBank mRNA | AK089483.1 | - | 15 - 16 | 2342 | 108 | |
AK149847.1 | - | 15 - 16 | 2305 | 108 | ||
AK154643.1 | - | 15 - 16 | 2316 | 108 | ||
AK161050.1 | - | 6 - 7 | 1245 | 108 | ||
AK171279.1 | - | 14 - 15 | 2462 | 108 | ||
AY156924.1 | - | 14 - 15 | 2186 | 108 | ||
BC128076.1 | - | 14 - 15 | 2186 | 40060132 | 108 | |
BC128077.1 | - | 14 - 15 | 2186 | 40060133 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |