Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hadha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178878.2 | NP_849209.1 | 17 - 18 | 1922 | 77 | |
GenBank mRNA | AK029017.1 | - | 17 - 18 | 1923 | 77 | |
AK150188.1 | - | 17 - 18 | 1920 | 77 | ||
AK170683.1 | - | 17 - 18 | 1923 | 77 | ||
AK203849.1 | - | 282 | 77 | |||
BC027156.1 | - | 6 - 7 | 817 | 77 | ||
BC037009.1 | - | 17 - 18 | 1910 | 5134605 | 77 | |
BC046978.1 | - | 17 - 18 | 1896 | 6315204 | 77 | |
BC058569.1 | - | 17 - 18 | 1918 | 6468514 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |