Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rftn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181397.2 | NP_852062.1 | 1 - 2 | 275 | 101 | |
GenBank mRNA | AK152130.1 | - | 1 - 2 | 197 | 101 | |
AK153288.1 | - | 1 - 2 | 197 | 101 | ||
AK155870.1 | - | 1 - 2 | 239 | 101 | ||
AK170913.1 | - | 1 - 2 | 239 | 101 | ||
AK172884.1 | - | 1 - 2 | 203 | 101 | ||
BC052074.1 | - | 1 - 2 | 275 | 6401192 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |