Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181820.2 | NP_861541.2 | 10 - 11 | 1532 | 88 | |
GenBank mRNA | AK050105.1 | - | 9 - 10 | 1427 | 88 | |
AY236493.1 | - | 10 - 11 | 1495 | 88 | ||
AY263162.1 | - | 10 - 11 | 1503 | 88 | ||
BC022758.1 | - | 2 - 3 | 222 | 88 | ||
BC040466.1 | - | 10 - 11 | 1444 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |