Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181820.2 | NP_861541.2 | 11 - 12 | 1718 | 84 | |
GenBank mRNA | AK050105.1 | - | 10 - 11 | 1613 | 84 | |
AY236493.1 | - | 11 - 12 | 1681 | 84 | ||
AY263162.1 | - | 11 - 12 | 1689 | 84 | ||
BC022758.1 | - | 3 - 4 | 408 | 84 | ||
BC040466.1 | - | 11 - 12 | 1630 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |