Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183020.1 | NP_898841.1 | 18 - 19 | 2553 | 77 | |
GenBank mRNA | AK155062.1 | - | 18 - 19 | 2600 | 77 | |
AK158626.1 | - | 18 - 19 | 2240 | 77 | ||
AK168745.1 | - | 18 - 19 | 2270 | 77 | ||
BC034083.1 | - | 12 - 13 | 1632 | 77 | ||
BC043451.1 | - | 13 - 14 | 1711 | 77 | ||
BC054483.1 | - | 18 - 19 | 2553 | 6391591 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |