Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183099.2 | NP_898922.2 | 1 - 2 | 457 | 77 | |
GenBank mRNA | AK007032.1 | - | 1 - 2 | 459 | 77 | |
BC114543.1 | - | 1 - 2 | 261 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |