Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183106.2 | NP_898929.2 | 23 - 24 | 3348 | 64 | |
GenBank mRNA | AK018641.1 | - | 7 - 8 | 988 | 64 | |
BC061503.1 | - | 13 - 14 | 1921 | 64 | ||
BC095993.1 | - | 8 - 9 | 1213 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |