Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183154.3 | NP_898977.2 | 4 - 5 | 1791 | 130 | |
GenBank mRNA | AK090334.1 | - | 5 - 6 | 1962 | 131 | |
AK129400.1 | - | 4 - 5 | 1766 | 130 | ||
AK139825.1 | - | 4 - 5 | 1812 | 130 | ||
AK170954.1 | - | 4 - 5 | 1792 | 130 | ||
BC050015.1 | - | 3 - 4 | 1212 | 30016571 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |