Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183154.3 | NP_898977.2 | 9 - 10 | 2402 | 75 | |
GenBank mRNA | AK090334.1 | - | 10 - 11 | 2573 | 75 | |
AK129400.1 | - | 9 - 10 | 2377 | 75 | ||
AK139825.1 | - | 9 - 10 | 2423 | 75 | ||
AK170954.1 | - | 9 - 10 | 2403 | 75 | ||
BC050015.1 | - | 8 - 9 | 1823 | 30016571 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |