Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183172.2 | NP_898995.1 | 9 - 10 | 1700 | 133 | |
GenBank mRNA | AY940666.1 | - | 9 - 10 | 1669 | 133 | |
BC051080.1 | - | 9 - 10 | 1697 | 6490136 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |