Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmiz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183208.3 | NP_899031.2 | 2 - 3 | 475 | 73 | |
GenBank mRNA | AK054366.1 | - | 2 - 3 | 369 | 73 | |
AK138053.1 | - | 2 - 3 | 475 | 73 | ||
AK210065.1 | - | 1 - 2 | 105 | 73 | ||
BC058646.1 | - | 2 - 3 | 393 | 6405777 | 73 | |
BC065120.1 | - | 2 - 3 | 368 | 6856060 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |