Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289768.1 | NP_001276697.1 | 1 - 2 | 134 | 67 | |
NM_183284.3 | NP_899107.1 | 1 - 2 | 134 | 67 | ||
GenBank mRNA | AK005709.1 | - | 1 - 2 | 117 | 67 | |
BC048619.1 | - | 1 - 2 | 101 | 67 | ||
BC087876.1 | - | 1 - 2 | 97 | 6773100 | 67 | |
DV068802.1 | - | 1 - 2 | 154 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |