Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY360471.1 | - | 531 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Inventoried |