Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010897.2 | NP_035027.1 | 28 - 29 | 3985 | 74 | |
GenBank mRNA | D30730.1 | - | 2 - 3 | 356 | 74 | |
D30731.1 | - | 2 - 3 | 356 | 74 | ||
L10370.1 | - | 27 - 28 | 3830 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |