Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010897.2 | NP_035027.1 | 5 - 6 | 693 | 138 | |
GenBank mRNA | L10370.1 | - | 4 - 5 | 538 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |