Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK034856.1 | - | 15 - 16 | 2211 | 63 | |
AK052103.1 | - | 13 - 14 | 1939 | 63 | ||
AK085336.1 | - | 14 - 15 | 2086 | 63 | ||
AK086462.1 | - | 15 - 16 | 2243 | 63 | ||
BC054120.1 | - | 15 - 16 | 2196 | 6506658 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |