Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK034856.1 | - | 6 - 7 | 845 | 90 | |
AK040344.1 | - | 5 - 6 | 741 | 90 | ||
AK085336.1 | - | 5 - 6 | 719 | 90 | ||
AK085362.1 | - | 6 - 7 | 884 | 90 | ||
AK086462.1 | - | 6 - 7 | 877 | 90 | ||
BC054120.1 | - | 6 - 7 | 830 | 6506658 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |