Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181564.2 | NP_853543.1 | 6 - 7 | 812 | 66 | |
GenBank mRNA | AB240192.1 | - | 6 - 7 | 812 | 66 | |
AY191584.1 | - | 6 - 7 | 812 | 66 | ||
AY227349.1 | - | 6 - 7 | 678 | 66 | ||
BC109009.2 | - | 6 - 7 | 812 | 66 | ||
BC150475.2 | - | 6 - 7 | 812 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |