Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163263.1 | NP_001156735.1 | 15 - 16 | 2640 | 70 | |
NM_182999.2 | NP_892044.1 | 16 - 17 | 2487 | 70 | ||
GenBank mRNA | AK220492.1 | - | 15 - 16 | 2556 | 70 | |
BC052482.1 | - | 16 - 17 | 2426 | 6308280 | 70 | |
BQ442966.1 | - | 3 - 4 | 402 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |