Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183017.2 | NP_898838.2 | 9 - 10 | 1372 | 66 | |
GenBank mRNA | AK084879.1 | - | 9 - 10 | 1339 | 66 | |
AK134345.1 | - | 9 - 10 | 1339 | 66 | ||
AK143496.1 | - | 8 - 9 | 1205 | 66 | ||
AK150116.1 | - | 9 - 10 | 1338 | 66 | ||
AM690756.1 | - | 9 - 10 | 1325 | 66 | ||
BC055368.1 | - | 9 - 10 | 1337 | 30055801 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |