Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slain1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198014.2 | NP_932131.2 | 5 - 6 | 1455 | 81 | |
GenBank mRNA | AK036190.1 | - | 7 - 8 | 1560 | 81 | |
BC025223.1 | - | 3 - 4 | 609 | 5010865 | 81 | |
BC079866.1 | - | 5 - 6 | 1455 | 6811096 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |