Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164559.1 | NP_001158031.1 | 13 - 14 | 1626 | 57 | |
NM_001164560.1 | NP_001158032.1 | 13 - 14 | 1550 | 57 | ||
NM_198020.2 | NP_932137.2 | 13 - 14 | 1653 | 57 | ||
GenBank mRNA | AK032945.1 | - | 11 - 12 | 3086 | 57 | |
AK084775.1 | - | 13 - 14 | 1593 | 57 | ||
AK155251.1 | - | 13 - 14 | 1619 | 57 | ||
AK159467.1 | - | 13 - 14 | 1591 | 57 | ||
AK170412.1 | - | 13 - 14 | 1515 | 57 | ||
AK171002.1 | - | 13 - 14 | 1618 | 57 | ||
AK212736.1 | - | 2 - 3 | 189 | 57 | ||
BB055133.2 | - | 1 - 2 | 181 | 57 | ||
BC027404.1 | - | 13 - 14 | 1622 | 4950980 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |