Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029985.2 | NP_084261.1 | 1 - 2 | 263 | 108 | |
GenBank mRNA | BC027203.1 | - | 1 - 2 | 195 | 3710091 | 108 |
BC031894.1 | - | 1 - 2 | 167 | 5362699 | 108 | |
CJ174174.1 | - | 1 - 2 | 264 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |