Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000001676.mm.1 |
NM_176837.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001676.mm.1 |
NM_176837.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001676.mm.1 |
TC1000000240.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000001673.mm.1 |
NM_176837.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001673.mm.1 |
NM_176837.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001673.mm.1 |
TC1000000240.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000001677.mm.1 |
NM_176837.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001677.mm.1 |
NM_176837.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001677.mm.1 |
TC1000000240.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000001678.mm.1 |
NM_176837.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001678.mm.1 |
NM_176837.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001678.mm.1 |
TC1000000240.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000001675.mm.1 |
NM_176837.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001675.mm.1 |
NM_176837.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001675.mm.1 |
TC1000000240.mm.1 |
MTA TC |