Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc23a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194333.3 | NP_919314.2 | 4 - 5 | 539 | 91 | |
GenBank mRNA | BC094893.1 | - | 4 - 5 | 543 | 30017276 | 91 |
BC117790.1 | - | 4 - 5 | 526 | 40092152 | 91 | |
BC117791.1 | - | 4 - 5 | 526 | 40092153 | 91 | |
U25739.1 | - | 4 - 5 | 551 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |