Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Accs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290782.1 | NP_001277711.1 | 3 - 4 | 527 | 110 | |
NM_183220.3 | NP_899043.1 | 3 - 4 | 582 | 110 | ||
GenBank mRNA | AK011867.1 | - | 3 - 4 | 431 | 110 | |
AK040509.1 | - | 3 - 4 | 424 | 110 | ||
BC039569.1 | - | 3 - 4 | 559 | 1449080 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |