Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027935.2 | NP_082211.1 | 2 - 3 | 132 | 68 | |
GenBank mRNA | AK014282.1 | - | 2 - 3 | 202 | 68 | |
AK147069.1 | - | 2 - 3 | 168 | 68 | ||
AK152630.1 | - | 2 - 3 | 216 | 68 | ||
AK152654.1 | - | 2 - 3 | 187 | 68 | ||
AK157214.1 | - | 2 - 3 | 205 | 68 | ||
AK164989.1 | - | 2 - 3 | 198 | 68 | ||
AK165636.1 | - | 2 - 3 | 181 | 68 | ||
BC094256.1 | - | 2 - 3 | 132 | 6820666 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |