Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dppa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139218.1 | NP_631964.1 | 1 - 2 | 182 | 87 | |
GenBank mRNA | AB072734.1 | - | 1 - 2 | 83 | 87 | |
AF490347.1 | - | 1 - 2 | 187 | 87 | ||
AK010723.1 | - | 1 - 2 | 87 | |||
AK135769.1 | - | 1 - 2 | 187 | 87 | ||
AK136263.1 | - | 1 - 2 | 187 | 87 | ||
AY082485.1 | - | 1 - 2 | 182 | 87 | ||
AY134859.1 | - | 1 - 2 | 194 | 87 | ||
BC099433.1 | - | 1 - 2 | 160 | 30933569 | 87 | |
BC100331.1 | - | 1 - 2 | 157 | 30934920 | 87 | |
BC107340.1 | - | 1 - 2 | 114 | 40055218 | 87 | |
BC107341.1 | - | 1 - 2 | 114 | 40055220 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |