Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145441.3 | NP_663416.1 | 5 - 6 | 617 | 106 | |
GenBank mRNA | AK031821.1 | - | 5 - 6 | 618 | 106 | |
AK041556.1 | - | 5 - 6 | 654 | 106 | ||
AK161940.1 | - | 5 - 6 | 758 | 106 | ||
BC004632.1 | - | 5 - 6 | 569 | 3585699 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |