Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100028040.mm.1 |
NM_001034097.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028040.mm.1 |
NM_001034097.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028040.mm.1 |
NM_001159503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028040.mm.1 |
NM_001159503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028040.mm.1 |
NM_011614.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028040.mm.1 |
NM_011614.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028040.mm.1 |
TC1100003059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028041.mm.1 |
NM_001034097.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028041.mm.1 |
NM_001034097.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028041.mm.1 |
NM_001159503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028041.mm.1 |
NM_001159503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028041.mm.1 |
NM_011614.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028041.mm.1 |
NM_011614.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028041.mm.1 |
TC1100003059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028045.mm.1 |
NM_001034097.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028045.mm.1 |
NM_001034097.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028045.mm.1 |
NM_001159503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028045.mm.1 |
NM_001159503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028045.mm.1 |
NM_011614.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028045.mm.1 |
NM_011614.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028045.mm.1 |
TC1100003059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028036.mm.1 |
NM_001034097.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028036.mm.1 |
NM_001034097.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028036.mm.1 |
NM_001159503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028036.mm.1 |
NM_001159503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028036.mm.1 |
NM_011614.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028036.mm.1 |
NM_011614.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028036.mm.1 |
TC1100003059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028037.mm.1 |
NM_001034097.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028037.mm.1 |
NM_001034097.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028037.mm.1 |
NM_001159503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028037.mm.1 |
NM_001159503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028037.mm.1 |
NM_011614.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028037.mm.1 |
NM_011614.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028037.mm.1 |
TC1100003059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028039.mm.1 |
NM_001034097.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028039.mm.1 |
NM_001034097.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028039.mm.1 |
NM_001159503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028039.mm.1 |
NM_001159503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028039.mm.1 |
NM_011614.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028039.mm.1 |
NM_011614.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028039.mm.1 |
TC1100003059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028047.mm.1 |
NM_001034097.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028047.mm.1 |
NM_001034097.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028047.mm.1 |
NM_001159503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028047.mm.1 |
NM_001159503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028047.mm.1 |
NM_011614.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028047.mm.1 |
NM_011614.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028047.mm.1 |
TC1100003059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028048.mm.1 |
NM_001034097.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028048.mm.1 |
NM_001034097.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028048.mm.1 |
NM_001159503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028048.mm.1 |
NM_001159503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028048.mm.1 |
NM_011614.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028048.mm.1 |
NM_011614.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028048.mm.1 |
TC1100003059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028038.mm.1 |
NM_001034097.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028038.mm.1 |
NM_001034097.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028038.mm.1 |
NM_001159503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028038.mm.1 |
NM_001159503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028038.mm.1 |
NM_011614.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028038.mm.1 |
NM_011614.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028038.mm.1 |
TC1100003059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028044.mm.1 |
NM_001034097.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028044.mm.1 |
NM_001034097.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028044.mm.1 |
NM_001159503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028044.mm.1 |
NM_001159503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028044.mm.1 |
NM_011614.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028044.mm.1 |
NM_011614.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028044.mm.1 |
TC1100003059.mm.1 |
MTA TC |