Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175638.3 | NP_783569.1 | 6 - 7 | 1571 | 104 | |
GenBank mRNA | AY184228.1 | - | 6 - 7 | 1540 | 104 | |
AY187027.1 | - | 6 - 7 | 1496 | 104 | ||
AY260059.1 | - | 6 - 7 | 1534 | 104 | ||
BC037114.1 | - | 6 - 7 | 1531 | 104 | ||
BC043677.1 | - | 6 - 7 | 944 | 104 | ||
BC096453.1 | - | 6 - 7 | 1527 | 4234311 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |