Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100009903.mm.1 |
NM_001277219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009903.mm.1 |
NM_001277221.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009903.mm.1 |
NM_133656.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009903.mm.1 |
TC1100001078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100009900.mm.1 |
NM_001277219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009900.mm.1 |
NM_001277221.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009900.mm.1 |
NM_133656.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009900.mm.1 |
TC1100001078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100009901.mm.1 |
NM_001277219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009901.mm.1 |
NM_001277221.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009901.mm.1 |
NM_133656.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009901.mm.1 |
TC1100001078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100009898.mm.1 |
NM_001277219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009898.mm.1 |
NM_001277221.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009898.mm.1 |
NM_133656.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009898.mm.1 |
TC1100001078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100009902.mm.1 |
NM_001277219.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009902.mm.1 |
NM_001277221.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009902.mm.1 |
NM_133656.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100009902.mm.1 |
TC1100001078.mm.1 |
MTA TC |