Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016870.3 | NP_058566.3 | 5 - 6 | 894 | 68 | |
NM_212441.2 | NP_997606.2 | 4 - 5 | 789 | 68 | ||
NM_212442.2 | NP_997607.2 | 4 - 5 | 692 | 68 | ||
GenBank mRNA | AB022340.2 | - | 4 - 5 | 680 | 68 | |
AK041060.1 | - | 4 - 5 | 664 | 68 | ||
AK077247.1 | - | 4 - 5 | 751 | 68 | ||
AK143946.1 | - | 692 | 68 | |||
AK165516.1 | - | 4 - 5 | 666 | 68 | ||
AY064696.1 | - | 4 - 5 | 765 | 68 | ||
BC015248.1 | - | 5 - 6 | 854 | 4239288 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |