Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253738.1 | NP_001240667.1 | 5 - 6 | 740 | 111 | |
NM_001271764.1 | NP_001258693.1 | 5 - 6 | 740 | 111 | ||
NM_022314.3 | NP_071709.3 | 6 - 7 | 723 | 111 | ||
GenBank mRNA | AK050665.1 | - | 5 - 6 | 586 | 111 | |
AK088111.1 | - | 5 - 6 | 600 | 111 | ||
AK088902.1 | - | 5 - 6 | 598 | 111 | ||
AK145238.1 | - | 5 - 6 | 597 | 111 | ||
AK145243.1 | - | 5 - 6 | 597 | 111 | ||
AK145804.1 | - | 5 - 6 | 597 | 111 | ||
AK146349.1 | - | 5 - 6 | 570 | 111 | ||
AK146557.1 | - | 5 - 6 | 722 | 111 | ||
AK151441.1 | - | 5 - 6 | 622 | 111 | ||
AK151500.1 | - | 5 - 6 | 597 | 111 | ||
AK151571.1 | - | 5 - 6 | 596 | 111 | ||
AK151578.1 | - | 5 - 6 | 597 | 111 | ||
AK151595.1 | - | 5 - 6 | 569 | 111 | ||
AK151602.1 | - | 5 - 6 | 597 | 111 | ||
AK151991.1 | - | 5 - 6 | 596 | 111 | ||
AK152260.1 | - | 5 - 6 | 596 | 111 | ||
AK152659.1 | - | 5 - 6 | 569 | 111 | ||
AK153031.1 | - | 5 - 6 | 569 | 111 | ||
AK160011.1 | - | 5 - 6 | 622 | 111 | ||
AK165428.1 | - | 5 - 6 | 596 | 111 | ||
AK166836.1 | - | 5 - 6 | 584 | 111 | ||
AK167268.1 | - | 5 - 6 | 574 | 111 | ||
AK167534.1 | - | 5 - 6 | 602 | 111 | ||
AK167581.1 | - | 5 - 6 | 597 | 111 | ||
AK168843.1 | - | 5 - 6 | 565 | 111 | ||
AK169102.1 | - | 5 - 6 | 568 | 111 | ||
AK169245.1 | - | 5 - 6 | 573 | 111 | ||
AK169821.1 | - | 5 - 6 | 614 | 111 | ||
BC029186.1 | - | 6 - 7 | 680 | 4502880 | 111 | |
BC055407.1 | - | 3866 | 111 | |||
BC092045.1 | - | 5 - 6 | 573 | 3591372 | 111 | |
X53753.1 | - | 5 - 6 | 608 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |