Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900012049.mm.1 |
NM_001205330.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012049.mm.1 |
NM_001205330.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012049.mm.1 |
TC0900001445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900012051.mm.1 |
NM_001205330.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012051.mm.1 |
NM_001205330.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012051.mm.1 |
NM_001205332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012051.mm.1 |
NM_001205332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012051.mm.1 |
NM_008633.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012051.mm.1 |
NM_008633.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012051.mm.1 |
TC0900001445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900012053.mm.1 |
NM_001205330.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012053.mm.1 |
NM_001205330.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012053.mm.1 |
NM_001205332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012053.mm.1 |
NM_001205332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012053.mm.1 |
NM_008633.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012053.mm.1 |
NM_008633.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012053.mm.1 |
TC0900001445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900012052.mm.1 |
NM_001205330.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012052.mm.1 |
NM_001205330.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012052.mm.1 |
NM_001205332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012052.mm.1 |
NM_001205332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012052.mm.1 |
NM_008633.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012052.mm.1 |
NM_008633.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012052.mm.1 |
TC0900001445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900012050.mm.1 |
NM_001205330.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012050.mm.1 |
NM_001205330.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012050.mm.1 |
NM_001205332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012050.mm.1 |
NM_001205332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012050.mm.1 |
NM_008633.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012050.mm.1 |
NM_008633.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012050.mm.1 |
TC0900001445.mm.1 |
MTA TC |