Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020277.2 | NP_064673.2 | 5 - 6 | 746 | 75 | |
GenBank mRNA | AB039952.1 | - | 4 - 5 | 663 | 75 | |
AF228681.1 | - | 5 - 6 | 746 | 75 | ||
AJ271092.2 | - | 4 - 5 | 659 | 75 | ||
AK035197.1 | - | 1 - 2 | 276 | 75 | ||
AK044908.1 | - | 1 - 2 | 361 | 75 | ||
AY280364.1 | - | 5 - 6 | 710 | 75 | ||
AY280365.1 | - | 5 - 6 | 710 | 75 | ||
BC133712.1 | - | 4 - 5 | 663 | 40143371 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |