Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083315.2 | NP_001076784.1 | 5 - 6 | 856 | 87 | |
NM_001289624.1 | NP_001276553.1 | 5 - 6 | 780 | 87 | ||
NM_001289625.1 | NP_001276554.1 | 5 - 6 | 780 | 87 | ||
NM_001289626.1 | NP_001276555.1 | 5 - 6 | 780 | 87 | ||
NM_001289627.1 | NP_001276556.1 | 5 - 6 | 514 | 87 | ||
NM_001289629.1 | NP_001276558.1 | 5 - 6 | 514 | 87 | ||
NM_022332.3 | NP_071727.2 | 5 - 6 | 856 | 87 | ||
GenBank mRNA | AF234884.1 | - | 5 - 6 | 807 | 87 | |
AF234885.1 | - | 5 - 6 | 807 | 87 | ||
AK079107.1 | - | 5 - 6 | 857 | 87 | ||
AK132103.1 | - | 5 - 6 | 810 | 87 | ||
AK151550.1 | - | 5 - 6 | 811 | 87 | ||
AK180967.1 | - | 1 - 2 | 126 | 87 | ||
AY007792.1 | - | 5 - 6 | 742 | 87 | ||
AY007793.1 | - | 5 - 6 | 742 | 87 | ||
AY007794.1 | - | 6 - 7 | 771 | 87 | ||
AY007795.1 | - | 6 - 7 | 771 | 87 | ||
AY007796.1 | - | 6 - 7 | 581 | 87 | ||
AY007797.1 | - | 6 - 7 | 581 | 87 | ||
AY007798.1 | - | 5 - 6 | 514 | 87 | ||
AY007799.1 | - | 5 - 6 | 514 | 87 | ||
AY551821.1 | - | 5 - 6 | 507 | 87 | ||
AY551822.1 | - | 5 - 6 | 507 | 87 | ||
BC024652.1 | - | 5 - 6 | 780 | 3660309 | 87 | |
BC051429.1 | - | 5 - 6 | 634 | 87 | ||
BC060630.1 | - | 5 - 6 | 697 | 87 | ||
BC108418.1 | - | 5 - 6 | 800 | 30603484 | 87 | |
CT010245.1 | - | 5 - 6 | 433 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |