Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290651.1 | NP_001277580.1 | 6 - 7 | 1180 | 120 | |
NM_001290652.1 | NP_001277581.1 | 6 - 7 | 1180 | 120 | ||
GenBank mRNA | AF285577.1 | - | 8 - 9 | 1006 | 120 | |
AK016533.1 | - | 9 - 10 | 1052 | 120 | ||
AK031490.1 | - | 6 - 7 | 1159 | 120 | ||
AK040521.1 | - | 6 - 7 | 1182 | 120 | ||
BC056382.1 | - | 6 - 7 | 870 | 6853936 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |