Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnksr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177751.2 | NP_808419.1 | 20 - 21 | 2959 | 81 | |
GenBank mRNA | AK122399.1 | - | 19 - 20 | 2883 | 81 | |
BC043093.1 | - | 20 - 21 | 2959 | 6405389 | 81 | |
BC060716.1 | - | 20 - 21 | 2959 | 6405389 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |