Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mospd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290523.1 | NP_001277452.1 | 4 - 5 | 474 | 126 | |
NM_001290524.1 | NP_001277453.1 | 4 - 5 | 474 | 126 | ||
NM_029730.4 | NP_084006.2 | 4 - 5 | 474 | 126 | ||
GenBank mRNA | AK010480.1 | - | 4 - 5 | 396 | 126 | |
AK031405.1 | - | 4 - 5 | 399 | 126 | ||
AK035985.1 | - | 4 - 5 | 434 | 126 | ||
AK039893.1 | - | 4 - 5 | 425 | 126 | ||
AK135188.1 | - | 4 - 5 | 429 | 126 | ||
AK156572.1 | - | 4 - 5 | 417 | 126 | ||
BC026425.1 | - | 4 - 5 | 400 | 4223175 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |