Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081116.1 | NP_001074585.1 | 3 - 4 | 3853 | 74 | |
GenBank mRNA | AK122250.1 | - | 3 - 4 | 508 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |