Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253892.1 | NP_001240821.1 | 15 - 16 | 2588 | 111 | |
NM_009207.3 | NP_033233.2 | 16 - 17 | 2784 | 111 | ||
GenBank mRNA | AK145930.1 | - | 16 - 17 | 2709 | 111 | |
AK147858.1 | - | 16 - 17 | 2740 | 111 | ||
AK190787.1 | - | 1 - 2 | 81 | 111 | ||
AK196438.1 | - | 1 - 2 | 81 | 111 | ||
AK200213.1 | - | 1 - 2 | 81 | 111 | ||
AK219574.1 | - | 1 - 2 | 81 | 111 | ||
BC002234.1 | - | 1 - 2 | 204 | 111 | ||
BC054102.1 | - | 15 - 16 | 2593 | 30017310 | 111 | |
J04036.1 | - | 16 - 17 | 2699 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |