Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000010725.mm.1 |
NM_001110218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010725.mm.1 |
NM_001110218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010725.mm.1 |
NM_176919.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010725.mm.1 |
NM_176919.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010725.mm.1 |
TC1000001512.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000010721.mm.1 |
NM_001110218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010721.mm.1 |
NM_001110218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010721.mm.1 |
NM_176919.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010721.mm.1 |
NM_176919.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010721.mm.1 |
TC1000001512.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000010724.mm.1 |
NM_001110218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010724.mm.1 |
NM_001110218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010724.mm.1 |
NM_176919.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010724.mm.1 |
NM_176919.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010724.mm.1 |
TC1000001512.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000010722.mm.1 |
NM_001110218.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010722.mm.1 |
NM_001110218.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010722.mm.1 |
NM_176919.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010722.mm.1 |
NM_176919.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000010722.mm.1 |
TC1000001512.mm.1 |
MTA TC |