Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081267.2 | NP_001074736.1 | 7 - 8 | 2730 | 82 | |
GenBank mRNA | AK032853.1 | - | 7 - 8 | 2706 | 82 | |
AK146675.1 | - | 7 - 8 | 2740 | 82 | ||
BC054454.1 | - | 2 - 3 | 221 | 82 | ||
BC085127.1 | - | 7 - 8 | 2731 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |