Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178414.3 | NP_848501.1 | 6 - 7 | 1103 | 64 | |
GenBank mRNA | AK132127.1 | - | 6 - 7 | 1104 | 64 | |
BC048390.1 | - | 6 - 7 | 1039 | 6312894 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |