Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026165.3 | NP_080441.1 | 2 - 3 | 411 | 75 | |
GenBank mRNA | AK004625.1 | - | 2 - 3 | 415 | 75 | |
AK008131.1 | - | 2 - 3 | 415 | 75 | ||
AK030346.1 | - | 2 - 3 | 412 | 75 | ||
AK031117.1 | - | 2 - 3 | 412 | 75 | ||
AK078383.1 | - | 421 | 75 | |||
AK147936.1 | - | 2 - 3 | 426 | 75 | ||
AK160834.1 | - | 2 - 3 | 412 | 75 | ||
BC020087.1 | - | 2 - 3 | 395 | 3990909 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |