Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038604.1 | NP_001033693.1 | 1 - 2 | 159 | 77 | |
NM_021364.2 | NP_067339.1 | 1 - 2 | 159 | 77 | ||
GenBank mRNA | AF139769.1 | - | 1 - 2 | 129 | 77 | |
AK036697.1 | - | 1 - 2 | 125 | 77 | ||
AK046600.1 | - | 1 - 2 | 146 | 77 | ||
AK137482.1 | - | 1 - 2 | 160 | 77 | ||
AK155409.1 | - | 1 - 2 | 176 | 77 | ||
BC104364.1 | - | 1 - 2 | 118 | 40049817 | 77 | |
BC104365.1 | - | 1 - 2 | 118 | 40049818 | 77 | |
BY205833.1 | - | 1 - 2 | 175 | 77 | ||
CN670295.1 | - | 1 - 2 | 121 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |